ENA245485736 Enamine

2,2,2-trifluoroethyl N-(2-chlorophenyl)carbamate

  • MDL number MFCD01013364
  • Cas Number : 1087798-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53145

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53145
Availability details for
Quantity
  • Availability
  • Add to Cart