ENA245485742 Enamine

N-hydroxy-3-phenoxypropanamide

  • MDL number MFCD02295792
  • Cas Number : 1016863-79-8
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53166
Availability details for
Quantity
  • Availability
  • Add to Cart