ENA245485751 Enamine

2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD03086210
  • Cas Number : 352018-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53177
Availability details for
Quantity
  • Availability
  • Add to Cart