ENA245485752 Enamine

1-(4-chlorophenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD03932876
  • Cas Number : 226881-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53179

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53179
Availability details for
Quantity
  • Availability
  • Add to Cart