ENA245485754 Enamine

(prop-2-en-1-yl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD12913217
  • Cas Number : 1197728-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53181
Availability details for
Quantity
  • Availability
  • Add to Cart