ENA245485759 Enamine

(3-{[ethyl(methyl)amino]methyl}phenyl)methanamine

  • MDL number MFCD11619508
  • Cas Number : 515131-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53186
Availability details for
Quantity
  • Availability
  • Add to Cart