ENA245485764 Enamine

1-(cyclopropylmethyl)piperidin-4-one

  • MDL number MFCD09937858
  • Cas Number : 49682-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53191
Availability details for
Quantity
  • Availability
  • Add to Cart