ENA245485774 Enamine

2-[(4-methoxyphenoxy)methyl]-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD11587924
  • Cas Number : 1082576-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53206
Availability details for
Quantity
  • Availability
  • Add to Cart