ENA245485791 Enamine

1-[2-(trifluoromethyl)phenoxy]propan-2-amine

  • MDL number MFCD06248883
  • Cas Number : 883542-94-7
  • Molecular Weight : 219.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[2-(trifluoromethyl)phenoxy]-2-propanamine
Inchi Code 1S/C10H12F3NO/c1-7(14)6-15-9-5-3-2-4-8(9)10(11,12)13/h2-5,7H,6,14H2,1H3
Inchi Key XRNYDNYFMQCOJD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53227
Availability details for
Quantity
  • Availability
  • Add to Cart