ENA245485792 Enamine

2-(chloromethyl)-5-cyclopropyl-1,3,4-oxadiazole

  • MDL number MFCD08059938
  • Cas Number : 915924-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53228
Availability details for
Quantity
  • Availability
  • Add to Cart