ENA245485793 Enamine

3-propoxybenzonitrile

  • MDL number MFCD09703612
  • Cas Number : 467462-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53229

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53229
Availability details for
Quantity
  • Availability
  • Add to Cart