ENA245485801 Enamine

1-benzyl-2,3-dihydro-1H-indole

  • MDL number MFCD12913229
  • Cas Number : 6037-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29...31
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53251

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53251
Availability details for
Quantity
  • Availability
  • Add to Cart