ENA245485817 Enamine

6-bromo-2-[2-(3-bromophenyl)ethenyl]quinoline-4-carboxylic acid

  • Cas Number : 926234-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53269
Availability details for
Quantity
  • Availability
  • Add to Cart