ENA245485818 Enamine

6-bromo-2-[2-(3-methoxyphenyl)ethenyl]quinoline-4-carboxylic acid

  • Cas Number : 926196-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53270
Availability details for
Quantity
  • Availability
  • Add to Cart