ENA245485820 Enamine

[3-(morpholin-4-yl)thiolan-3-yl]methanamine

  • MDL number MFCD09933042
  • Cas Number : 933749-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53272
Availability details for
Quantity
  • Availability
  • Add to Cart