ENA245485822 Enamine

[1-(2-methoxyphenyl)cyclopentyl]methanamine

  • MDL number MFCD09261728
  • Cas Number : 927993-53-1
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(2-methoxyphenyl)cyclopentyl]methanamine
Inchi Code 1S/C13H19NO/c1-15-12-7-3-2-6-11(12)13(10-14)8-4-5-9-13/h2-3,6-7H,4-5,8-10,14H2,1H3
Inchi Key SDHQESURORPKBB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53274

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-53274
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart