ENA245485825 Enamine

2-[2-(1H-imidazol-1-yl)ethoxy]aniline

  • MDL number MFCD11652479
  • Cas Number : 464913-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53277

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53277
Availability details for
Quantity
  • Availability
  • Add to Cart