ENA245485826 Enamine

2-(chloromethyl)-5-methoxy-1,3,4-thiadiazole

  • MDL number MFCD12913232
  • Cas Number : 3914-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-40
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53278
Availability details for
Quantity
  • Availability
  • Add to Cart