ENA245485829 Enamine

4-methyl-2-phenyl-1-(piperidin-4-yl)piperazine trihydrochloride

  • MDL number MFCD12913233
  • Cas Number : 1211321-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53284
Availability details for
Quantity
  • Availability
  • Add to Cart