ENA245485831 Enamine

2-(6-chloro-1H-indol-1-yl)acetic acid

  • MDL number MFCD11212282
  • Cas Number : 943654-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53286

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53286
Availability details for
Quantity
  • Availability
  • Add to Cart