ENA245485838 Enamine

1-(2-nitrophenyl)-3-(prop-2-yn-1-yl)urea

  • MDL number MFCD12913236
  • Cas Number : 1197861-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53296
Availability details for
Quantity
  • Availability
  • Add to Cart