ENA245485841 Enamine

3-(2-bromo-4-chlorophenoxy)propanoic acid

  • MDL number MFCD09044366
  • Cas Number : 926217-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53302
Availability details for
Quantity
  • Availability
  • Add to Cart