ENA245485842 Enamine

3-[4-(propan-2-yl)phenoxy]propanoic acid

  • MDL number MFCD09049892
  • Cas Number : 926257-24-1
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53304
Availability details for
Quantity
  • Availability
  • Add to Cart