ENA245485847 Enamine

(6-bromo-2H-1,3-benzodioxol-5-yl)methanol

  • MDL number MFCD02082027
  • Cas Number : 6642-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53315

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53315
Availability details for
Quantity
  • Availability
  • Add to Cart