ENA245485848 Enamine

1-(2,3-dihydro-1H-inden-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD13195868
  • Cas Number : 1009-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53317
Availability details for
Quantity
  • Availability
  • Add to Cart