ENA245485854 Enamine

6-bromo-2-[(E)-2-[4-(trifluoromethoxy)phenyl]ethenyl]quinoline-4-carboxylic acid

  • MDL number MFCD09048457
  • Cas Number : 926199-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 311...313
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53323
Availability details for
Quantity
  • Availability
  • Add to Cart