ENA245485860 Enamine

3-cyclopropylthiophene-2-carboxylic acid

  • MDL number MFCD13195793
  • Cas Number : 29488-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22174
Availability details for
Quantity
  • Availability
  • Add to Cart