ENA245485861 Enamine

2,2,2-trifluoroethyl N-(1,3-thiazol-2-yl)carbamate

  • MDL number MFCD13195871
  • Cas Number : 1197741-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53333
Availability details for
Quantity
  • Availability
  • Add to Cart