ENA245485863 Enamine

methyl 4-chloro-5-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD13195873
  • Cas Number : 1281872-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53335
Availability details for
Quantity
  • Availability
  • Add to Cart