ENA245485875 Enamine

3-(2,5-difluorophenyl)propanoic acid

  • MDL number MFCD04116053
  • Cas Number : 130408-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53351
Availability details for
Quantity
  • Availability
  • Add to Cart