ENA245485876 Enamine

2,2-dimethylpropanecarbonyl isothiocyanate

  • MDL number MFCD09734773
  • Cas Number : 93700-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22223
Availability details for
Quantity
  • Availability
  • Add to Cart