ENA245485877 Enamine

2-[(prop-2-en-1-yl)amino]acetic acid hydrochloride

  • MDL number MFCD08061162
  • Cas Number : 3182-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53353
Availability details for
Quantity
  • Availability
  • Add to Cart