ENA245485880 Enamine

8-bromo-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD09744046
  • Cas Number : 204377-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53358
Availability details for
Quantity
  • Availability
  • Add to Cart