ENA245485881 Enamine

3-(1,3-dioxo-octahydro-1H-isoindol-2-yl)-4-methylbenzoic acid

  • MDL number MFCD01576789
  • Cas Number : 895766-35-5
  • Molecular Weight : 287.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1,3-dioxooctahydro-2H-isoindol-2-yl)-4-methylbenzoic acid
Inchi Code 1S/C16H17NO4/c1-9-6-7-10(16(20)21)8-13(9)17-14(18)11-4-2-3-5-12(11)15(17)19/h6-8,11-12H,2-5H2,1H3,(H,20,21)
Inchi Key HFSSAWJPQSZVAL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53359
Availability details for
Quantity
  • Availability
  • Add to Cart