ENA245485885 Enamine

4-(2-methoxyethoxy)-2-(trifluoromethyl)aniline

  • MDL number MFCD08688042
  • Cas Number : 946784-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53365
Availability details for
Quantity
  • Availability
  • Add to Cart