ENA245485903 Enamine

2-(4-chlorobenzenesulfonyl)ethan-1-amine hydrochloride

  • MDL number MFCD03840166
  • Cas Number : 85052-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-219
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53396
Availability details for
Quantity
  • Availability
  • Add to Cart