ENA245485911 Enamine

3-amino-1,1,1-trifluoro-2-(1-methyl-1H-imidazol-2-yl)propan-2-ol dihydrochloride

  • MDL number MFCD13195884
  • Cas Number : 1197881-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53416
Availability details for
Quantity
  • Availability
  • Add to Cart