ENA245485914 Enamine

2-(thiophen-3-yl)-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD12818487
  • Cas Number : 1184003-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53424
Availability details for
Quantity
  • Availability
  • Add to Cart