ENA245485921 Enamine

N-(2,2,2-trifluoroethyl)pyridin-2-amine

  • MDL number MFCD11124025
  • Cas Number : 77262-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53432
Availability details for
Quantity
  • Availability
  • Add to Cart