ENA245485943 Enamine

1-(3,4-dimethylphenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09815095
  • Cas Number : 1017417-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53482
Availability details for
Quantity
  • Availability
  • Add to Cart