ENA245485947 Enamine

3-ethyl-1-hydroxycyclopentane-1-carboxylic acid

  • MDL number MFCD11181324
  • Cas Number : 1094307-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53491
Availability details for
Quantity
  • Availability
  • Add to Cart