ENA245485952 Enamine

1-[(2E)-3-(4-chlorophenyl)prop-2-enoyl]piperidine-2-carboxylic acid

  • MDL number MFCD09934104
  • Cas Number : 1103961-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53498

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53498
Availability details for
Quantity
  • Availability
  • Add to Cart