ENA245485953 Enamine

2-[4-(pyridin-2-yl)-1,3-thiazol-2-yl]acetonitrile

  • MDL number MFCD09938083
  • Cas Number : 1017049-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53499
Availability details for
Quantity
  • Availability
  • Add to Cart