ENA245485956 Enamine

4-(6-methoxynaphthalen-2-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD11217988
  • Cas Number : 1096308-30-3
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53502
Availability details for
Quantity
  • Availability
  • Add to Cart