ENA245485960 Enamine

2-(4-methyl-1,4-diazepan-1-yl)cyclopentan-1-amine

  • MDL number MFCD11172959
  • Cas Number : 1042646-03-6
  • Molecular Weight : 197.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53508
Availability details for
Quantity
  • Availability
  • Add to Cart