ENA245485963 Enamine

4-(pentafluoroethoxy)benzoic acid

  • MDL number MFCD11858215
  • Cas Number : 79865-59-1
  • Molecular Weight : 256.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1,1,2,2,2-pentafluoroethoxy)benzoic acid
Inchi Code 1S/C9H5F5O3/c10-8(11,12)9(13,14)17-6-3-1-5(2-4-6)7(15)16/h1-4H,(H,15,16)
Inchi Key SUTOUQPTCJCIKM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53512

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-53512
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart