ENA245485976 Enamine

1-[3-(4-carbamoylphenyl)prop-2-enoyl]pyrrolidine-2-carboxylic acid

  • Cas Number : 1485808-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53539
Availability details for
Quantity
  • Availability
  • Add to Cart