ENA245485981 Enamine

6-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD06203633
  • Cas Number : 209920-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53545
Availability details for
Quantity
  • Availability
  • Add to Cart