ENA245485985 Enamine

6-bromo-2-{2-[4-(dimethylamino)phenyl]ethenyl}quinoline-4-carboxylic acid

  • Cas Number : 859803-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53549
Availability details for
Quantity
  • Availability
  • Add to Cart