ENA245485993 Enamine

2-(2-amino-4-phenyl-1,3-thiazol-5-yl)acetic acid

  • MDL number MFCD00022453
  • Cas Number : 49779-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53566
Availability details for
Quantity
  • Availability
  • Add to Cart